Learn More
Isonicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals
£66.10 - £447.00
Chemical Identifiers
CAS | 39178-35-3 |
---|---|
Molecular Formula | C6H5Cl2NO |
Molecular Weight (g/mol) | 178.01 |
MDL Number | MFCD00012830 |
InChI Key | BNTRVUUJBGBGLZ-UHFFFAOYSA-N |
Synonym | isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl |
PubChem CID | 12262826 |
IUPAC Name | pyridine-4-carbonyl chloride;hydrochloride |
SMILES | [H+].[Cl-].ClC(=O)C1=CC=NC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10012680
|
Thermo Scientific Acros
211680250 |
25 g | Plastic bottle |
£66.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10762251
|
Thermo Scientific Acros
211681000 |
100 g | Plastic bottle |
£194.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10043813
|
Thermo Scientific Acros
211682500 |
250 g | Plastic bottle |
£447.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
39178-35-3 | |
178.01 | |
BNTRVUUJBGBGLZ-UHFFFAOYSA-N | |
12262826 | |
[H+].[Cl-].ClC(=O)C1=CC=NC=C1 |
C6H5Cl2NO | |
MFCD00012830 | |
isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl | |
pyridine-4-carbonyl chloride;hydrochloride |
Specifications
39178-35-3 | |
Beige to White | |
96% min. (Argentometry) | |
MFCD00012830 | |
isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl | |
[H+].[Cl-].ClC(=O)C1=CC=NC=C1 | |
178.01 | |
178.02 | |
Crystalline Powder |
153.0°C to 161.0°C | |
Authentic | |
C6H5Cl2NO | |
22, 46 | |
BNTRVUUJBGBGLZ-UHFFFAOYSA-N | |
pyridine-4-carbonyl chloride;hydrochloride | |
12262826 | |
97% | |
Isonicotinoyl chloride hydrochloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 254-331-2
RUO – Research Use Only