missing translation for 'onlineSavingsMsg'
Learn More

Iodic acid, ACS, 99.5% min, Thermo Scientific Chemicals

£46.63 - £190.95

Chemical Identifiers

CAS 7782-68-5
Molecular Formula HIO3
Molecular Weight (g/mol) 175.909
MDL Number MFCD00011348
InChI Key ICIWUVCWSCSTAQ-UHFFFAOYSA-N
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name iodic acid
SMILES OI(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355019
View Documents Promotion Details
Thermo Scientific Alfa Aesar
087681.18
50 g
£46.63
50g
Please to purchase this item. Need a web account? Register with us today!
11345019
View Documents Promotion Details
Thermo Scientific Alfa Aesar
087681.30
250 g
£190.95
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-68-5
175.909
ICIWUVCWSCSTAQ-UHFFFAOYSA-N
24345
iodic acid
HIO3
MFCD00011348
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
CHEBI:24857
OI(=O)=O

Specifications

Powder/chunks
110°C (decomposition)
Light sensitive
HIO3
MFCD00011348
Soluble in water and methanol.
OI(=O)=O
175.909
CHEBI:24857
ACS
7782-68-5
4.629 g/cm3
99.5%
Powder/Chunks
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
ICIWUVCWSCSTAQ-UHFFFAOYSA-N
iodic acid
24345
175.91
Iodic Acid
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-962-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Iodic acid, ACS

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Iodic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only