missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ GenoType Tsp DNA Polymerase

Platinum™ GenoType Tsp DNA Polymerase is designed and qualified specifically for amplification of dinucleotide repeat markers in PCR-based genotyping applications.

Brand:  Invitrogen™ 11448032

Code : 50

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 10694793

  • £2940.00 / Each
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Platinum™ GenoType Tsp DNA Polymerase is designed and qualified specifically for amplification of dinucleotide repeat markers in PCR-based genotyping applications. The enzyme is pre-complexed with a thermolabile inhibitor containing monoclonal antibodies to Tsp DNA polymerase, providing an automatic 'hot-start' method that improves PCR specificity and allows for room temperature reaction assembly.

Platinum™ GenoType Tsp DNA Polymerase:
• Exhibits minimal non-templated nucleotide addition to PCR products
• Lacks both 5´ and 3´ exonuclease activity
• Amplifies fragments up to 500 bp

Application
Amplification of dinucleotide repeat loci in applications in which elimination of non-templated nucleotide addition is desirable.

Unit definition
One unit of Platinum™ GenoType Tsp DNA Polymerase has been functionally determined to be equivalent to one unit of Taq DNA Polymerase in amplification of dinucleotide repeats using standard Taq DNA polymerase reaction conditions. One Tsp DNA polymerase unit approximates 2.5 activity units. An activity unit incorporates 10 nmoles of deoxyribonucleotide into acid-precipitable material in 30 min at 74°C under optimized reaction conditions.

Specifications

Specifications

• Platinum GenoType Tsp DNA Polymerase (500 μL at 5 Tsp U/μL)
• 10X PCR Buffer (10 x 1.25 mL)
• 50 mM MgCl2 (5 x 1 mL)

Store at -20°C. Guaranteed stable for 6 months when properly stored.
Tsp Genotyping Polymerase
Wet or Dry Ice
1X
2000 Reactions
Separate Components
500 μL
Low
Standard
Hot-start PCR
Built-In Hot Start
Blunt
500 bp or less
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.