missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ EX Agarose Gels, 1%
Precast, fast-resolving gels with 11 wells and an openable format
£128.00 - £261.00
Specifications
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder, Millennium™ RNA Markers |
---|---|
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | EX Agarose Gel |
Sample Loading Volume | 20 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11571186
|
Invitrogen™
G401001 |
10 Gels/Pk. |
£128.00
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17176448
|
Invitrogen™
G402021 |
20 Gels/Pk. |
£261.00
Pack of 20 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
- Contain a proprietary fluorescent nucleic acid stain (excitation: 490nm, emission: 520nm) that can be viewed with blue light, which minimizes DNA damage and allows detection down to 1ng/band of DNA
- Complete separation (by molecular weight) in 10 minutes
- Live monitoring of DNA migration without the hazards of UV light
- Easy access to the gel for extraction of DNA bands or transfer to membranes for Southern blotting
- Demonstrate over five-fold greater sensitivity than comparable gels containing ethidium bromide allows less sample use per run
- Fast analysis of RNA samples to check on their integrity before proceeding with downstream application
- The gel cassette is designed to open with a Gel Knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis
- Single comb well format
- Sample Type: Double-Stranded DNA (dsDNA), Single-Stranded RNA
- Sample Volume; 20μL
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
E-Gel™ 1 Kb Plus Express Ladder, Millennium™ RNA Markers | |
E-Gel | |
20 μL | |
SYBR™ Gold | |
Molecular Weight | |
Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA | |
Single Comb |
1% | |
EX Agarose Gel | |
100 to 5000 bp | |
11-well | |
Preparative without dyes, Non-preparative | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.