missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Inlet Liners for Basic Organic Analysis for Agilent™ GCs: Split Liner

Specifically designed to ensure a highly inert GC pathway for basic compounds

Brand:  Restek™ 20783-211.25

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12922799

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Split liner design with base-deactivated wool packing material.

Base-deactivation process:

  • Bonds basic functional groups to the analytical surface to reduce adsorption and priming effects
  • I.D. 4mm
  • Improves peak shape, response and reproducibility when compared to standard deactivation

  • Base-deactivated wool prevents the vaporized sample from contacting the adsorptive hot metal inlet disc surface

Specifications

Specifications

Base Deactivated Wool
25/Pk.
0.4 cm
Agilent GCs Equipped with Split/Splitless Inlets
Split Base Deactivated Straight Inlet Liner
7.85 cm
0.63 cm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers