missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Cool pH Electrodes
A glass pH electrode with an integrated temperature sensor rated to -30°C, making it ideal for measuring cold samples.
£307.00 - £414.00
Specifications
Junction | Immovable Glass Sleeve |
---|---|
Material | Glass |
pH Range | 1 to 11 |
Membrane | LoT Glass |
Temperature (Metric) Range | -30°C to +80°C |
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
10642244
|
METTLER TOLEDO™
51343174 |
InLab™ Cool | Combination Electrode | S7 | S7 to BNC (30281915) |
£307.00
Each |
In Stock
Log in to see stock. |
||
15539601
|
METTLER TOLEDO™
30247850 |
InLab™ Cool Pro-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | Multi-Pin | Multi-Pin to BNC/RCA (30281896) |
£414.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Cold samples: can measure down to -30°C
- TRIS compatible
Accessories & Care
- Immovable glass sleeve junction, refillable with FRISCOLYT-B™ (51343185)
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Immovable Glass Sleeve | |
1 to 11 | |
-30°C to +80°C |
Glass | |
LoT Glass |