missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(III) selenide, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Indium(III) selenide is used in photovoltaic devices. It is also used for optoelectronics and in solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽890°C |
Color | Silver to Gray |
Physical Form | Pieces |
Quantity | 5 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 466.52 |
Odor | Odorless |
Appearance | Silver gray pieces |
Density | 5.67 g/cm3 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'