missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ In-flow Safety Cans
For use in storing reagents in approved containers when drawing solvents into HPLC machines
£502.88
Specifications
Product Type | Safety Can |
---|---|
Certifications/Compliance | FM, NFPA, OSHA |
Color | White |
Material | Polyethylene |
Includes | Self Closing Fill/Pour Spout |
Product Code | Brand | Height (English) | Height (Metric) | Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Height (Metric) | Shape | Price | Quantity & Availability | |||
11909405
|
Justrite™
12164 |
9.5 in. | 24.1 cm | Oval | N/A | ||||
Description
- Made of high density polyethylene
- Equipped with a required self-closing fill/pour spout with stainless steel flame arrester
- Two quick-disconnect fittings: one polypropylene for venting, and one stainless steel to handle the solvent in-flow
- Stainless steel disconnect includes 1/8 in. OD (1/16 in.ID) and 1/16 in. OD (1/32 in. ID) compression fittings
Specifications
Safety Can | |
White | |
Self Closing Fill/Pour Spout |
FM, NFPA, OSHA | |
Polyethylene |