Learn More
Invitrogen™ IL-12 (IL-12A) Porcine ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ ESIL12AX5
Additional Details : Weight : 0.33800kg
Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.
Description
The Porcine Interleukin 12 (IL-12A) ELISA quantitates Porc IL-12A in porcine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Porc IL-12A. Principle of the method The Porcine IL-12A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Interleukin-12 (IL-12) is a heterodimeric 70 kD (p70) cytokine composed of two covalently linked, glycosylated chains, 40kD (p40) encoded by the IL12B gene and 35-kD (p35) encoded by the IL12A gene. IL-12 is mainly produced by monocytes, macrophages, and dendritic cells in response to bacterial products such as lipopolysaccharides (LPS), to intracellular pathogens or upon interaction with activated T cells. IL-12 was originally discovered because of its ability to induce interferon-gamma (IFN-γ) production, cell proliferation, and cytotoxicity mediated by natural killer cells and T cells. It is now established that IL-12 also plays a key role in the development of Th1 responses, leading to IFN-γ and IL-2 production. These cytokines can in turn promote T-cell responses and macrophage activation.Specifications
Q28938,Q29053 | |
2.2 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
Colorimetric Microplate Reader | |
IL12A, IL12B | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
Porcine | |
4 hr. 45 min. |
2.458 pg/mL to 600 pg/mL | |
2.2 ng/ml to 600 ng/ml | |
Biotin | |
Cytokines & Receptors | |
Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: porcine IL-1 beta, IL-4, IL-6, IL-8, IL-10, IL-13, GM-CSF, IFN-gamma, TGF beta 1, TNF alpha | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
397053, 397076 | |
<12% | |
Stop Solution, Biotinylated Detection Antibody, SAV-HRP, Assay Diluent concentrate, Chromogen, Standard, Pre-coated 96 well plate, Wash Buffer, Adhesive Plate Covers | |
5 x 96 Tests | |
Oncology | |
2°C to 8°C | |
1 hr. 20 min. |