missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ICOS (CD278) Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHICOSX5

Additional Details : Weight : 0.33820kg

 View more versions of this product

Product Code. 17105469

  • £2320.00 / Each
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

The Human ICOS (CD278) ELISA quantitates Hu ICOS in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ICOS. Principle of the method The Human ICOS solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CD278 (ICOS) belongs to the CD28 and CTLA-4 cell-surface receptor family, forms homodimers, and is highly expressed in activated T-cells. CD278 is essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. CD278 enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells. Diseases associated with CD278 dysfunction include Common Variable 1 Immunodeficiency and ICOS-related Common Related Immunodeficiency.
Specifications

Specifications

Q9Y6W8
1.2 ng/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
5 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
29851
<12%
Pre-coated 96 well plates, Wash Buffer, Biotinylated Detection Antibody, Adhesive Plate Covers, Chromogen, SAV-HRP, Assay Diluent Concentrate, Stop Solution, Standard
5 x 96 Tests
Extracellular Matrix & Cell Adhesion
2°C to 8°C
Human
4 hr. 45 min.
1.229 ng/mL to 300 ng/mL
1.2 ng/mL to 300 ng/mL
Biotin
Cell Structure & Organelle Proteins
This ELISA pair antibody detects human ICOS. Other species not determined.
Human
Colorimetric Microplate Reader
AILIM,CD278,CVID1
<10%
HRP
RUO
Plasma, 25 μ;L; Serum, 25 μ;L; Supernatant, 100 μ;L
AILIM, CD278, CVID1
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers