missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Sulphite for Ion Chromatography 1g/L H2O matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£49.50 - £119.00
Specifications
Physical Form | Liquid |
---|---|
Concentration | 1 g/L |
Type | Single-component Standard Solution |
Solvent or Matrix | H2O |
Chemical Name or Material | Hydrogen Sulphite |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12962225
![]() |
Fisher Chemical
A 053 100 |
100 mL | Bottle |
£49.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12972225
![]() |
Fisher Chemical
A 053 150 |
250 mL | Bottle |
£70.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12982225
![]() |
Fisher Chemical
A 053 500 |
500 mL | Bottle |
£119.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Liquid | |
Single-component Standard Solution | |
Hydrogen Sulphite |
1 g/L | |
H2O |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrogen Sulphite for Ion Chromatography 1g/L H2O matrix, Fisher chemical™