missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide 35%, J.T. Baker™

Product Code. 10688331
Click to view available options
1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Solutions for Chemical Testing

Specifications

CAS 7722-84-1
Boiling Point ∼107°C (1013 hPa)
Melting Point <0°C
Assay Percent Range min. 35%
Molecular Formula H2O2
MDL Number MFCD00011333
Density 1.11 g/cm³ (20°C)
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name peroxol
Molecular Weight (g/mol) 34.01
Packaging Plastic bottle
Quantity 1 L
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'