missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, Technical grade, Honeywell Fluka™

Catalog Number p-7071690
Click to view available options
Quantity:
1 L
5 L
6 x 1 L
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7664-39-3
FH
20.01
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
14917
CHEBI:29228
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7664-39-3
FH
20.01
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
14917
CHEBI:29228

Technical, 40-45%

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY
Concentration 40 to 45%
Density 1.15g/mL at 25°C
Vapor Pressure 25mmHg (20°C)
Quantity 1 L
UN Number 1790
SMILES F
IUPAC Name fluorane
Formula Weight 20.01g/mol
Grade Technical
Chemical Name or Material Hydrofluoric Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'