missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 33% to 36%, ULTREX, J.T. Baker™

Product Code. 10782232
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

pH <1
Color Colorless
CAS 7647-01-0
Melting Point 0°C
Boiling Point 50°C
Density 1.2g/cm3
Odor Pungent
Assay Percent Range 33% to 36%
Quantity 500 mL
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
UN Number 1789
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
Solubility Information Fully miscible in water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46
Grade ULTREX™
Chemical Name or Material Hydrochloric Acid
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'