Learn More
HUS1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565122
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: GASSLKLQLTH KRRPSLTVAV ELVSSLGRAR SVVHDLPVRV LPRRVWRDCL PPSLRASDAS IRLPRWRTLR SIVERMANVG SHVL Highest antigen sequence identity to the following orthologs - mouse 67%, rat 66%.
HUS1Bis most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. HUS1B can interact with the check point protein RAD1 but not with RAD9. Overexpression of HUS1B has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1. The protein encoded by this gene is most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. This protein can interact with the check point protein RAD1 but not with RAD9. Overexpression of this protein has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1.Specifications
HUS1B | |
Polyclonal | |
Unconjugated | |
Hus1b | |
Checkpoint protein HUS1B; hHUS1B; HUS1 checkpoint clamp component B; HUS1 checkpoint homolog b; HUS1 checkpoint homolog b (S. pombe); Hus1 homolog b (S. pombe); Hus1b; MGC126746; MGC126748; mHUS1B; RP11-532F6.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
135458 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NHY5 | |
Hus1b | |
Recombinant Human HUS1B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only