Learn More
Abnova™ Human PAK4 Partial ORF (AAH02921, 68 a.a. - 157 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
£280.00 - £425.00
Specifications
Accession Number | AAH02921 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10298 |
Molecular Weight (g/mol) | 35.53kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16194852
|
Abnova™
H00010298-Q01.10UG |
10 ug |
£280.00
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16104862
|
Abnova™
H00010298-Q01.25UG |
25 ug |
£425.00
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. They serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK4 interacts specifically with the GTP-bound form of Cdc42Hs and weakly activates the JNK family of MAP kinases. PAK4 is a mediator of filopodia formation and may play a role in the reorganization of the actin cytoskeleton. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: KTIVRGSKGAKDGALTLLLDEFENMSVTRSNSLRRDSPPPPARARQENGMPEKPPGPRSPQREPQRVSHEQFRAALQLVVDPGDPRSYLDSpecifications
AAH02921 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAK4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10298 | |
PAK4 (Human) Recombinant Protein (Q01) | |
KTIVRGSKGAKDGALTLLLDEFENMSVTRSNSLRRDSPPPPARARQENGMPEKPPGPRSPQREPQRVSHEQFRAALQLVVDPGDPRSYLD | |
RUO | |
PAK4 | |
Recombinant | |
wheat germ expression system |