Learn More
Abnova™ Human LEPRE1 Partial ORF (AAH15309, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064175-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an enzyme that is a member of the collagen prolyl hydroxylase family. These enzymes are localized to the endoplasmic reticulum and their activity is required for proper collagen synthesis and assembly. Mutations in this gene are associated with osteogenesis imperfecta type VIII. Two alternatively spliced transcript variants encoding different isoforms have been described. Other variants may exist, but their biological validity has not been determined. [provided by RefSeq]
Sequence: DPRVREVMNQNLAYYAAMLGEEHTRSIGPRESAKEYRQRSLLEKELLFFAYDVFGIPFVDPDSWTPEEVIPKRLQEKQKSERETAVRISQEIGNLMKEIESpecifications
AAH15309 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DPRVREVMNQNLAYYAAMLGEEHTRSIGPRESAKEYRQRSLLEKELLFFAYDVFGIPFVDPDSWTPEEVIPKRLQEKQKSERETAVRISQEIGNLMKEIE | |
RUO | |
LEPRE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64175 | |
LEPRE1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GROS1/MGC117314/P3H1 | |
LEPRE1 | |
Recombinant | |
wheat germ expression system |