missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-17 E ELISA Kit
Human IL-17 E quantitates human IL-17 E in serum, plasma, supernatant.
Brand: Invitrogen™ EH260RB
This item is not returnable.
View return policy
Description
Human IL-17 E quantitates human IL-17 E in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-17 E.
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.Specifications
il 17 e; IL17E; IL-17 e; IL25; IL-25; ILN; Interleukin; interleukin 17 E; interleukin 25; Interleukin-17 E; interleukin-25; RGD1561632; UNQ3120/PRO10272 | |
0.4 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
IL25 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.4 to 100 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q9H293 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
IL-25 | |
4 hr 45 min |