Learn More
Invitrogen™ IgE Human Uncoated ELISA Kit with Plates
For quantitative detection of human IgE.
£174.00 - £604.00
Specifications
Target Species | Human |
---|---|
For Use With (Application) | ELISA |
Synonym | Immunoglobulin E |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15520957
|
Invitrogen™
88-50610-76 |
20 x 96 Tests |
£604.00
Pack of 20 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
15510957
|
Invitrogen™
88-50610-22 |
2 x 96 Tests |
£174.00
Pack of 2 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15540957
|
Invitrogen™
88-50610-86 |
10 x 96 Tests |
£392.00
Pack of 10 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
The Human Immunoglobulin E (Hu IgE) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IgE. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.Specifications
Human | |
ELISA | |
Immunoglobulin E |
For Research Use Only