missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Heat Stable bFGF Recombinant Protein
Gibco Heat Stable Recombinant Human Bfgf is a heat stable version of basic FGF (Fibroblast Growth Factor) engineered for greater stability in cell culture conditions. HS bFGF maintains at least 50% more activity after 72 hours, in cell culture conditions than competitor product.
Brand: Gibco™ PHG0369
This item is not returnable.
View return policy
Description
Human Heat Stable bFGF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
- Superior performance - Retains bioactivity without the use of artificially high concentrations.
- Easy to use - Direct replacement for your current bFGF allowing for easy substitution in protocols.
- Advancement without compromise - > 90% sequence homology to native bFGF, leaving heparin and FGF receptor binding sites untouched
Specifications
Cell Culture | |
100 μg | |
155 aa + 20 aa for N-terminal tag for purification purposes | |
Heat Stable Recombinant Human bFGF | |
Sterile | |
Carrier-Free | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors and Receptors | |
Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis |
19 kDa | |
Less than 1 EU/ug | |
ED50 range = 0.6-1.1 ng⁄mL | |
Human | |
Store the lyophilized protein at -20°C, preferably desiccated. | |
< 1 EU/μg | |
E. coli | |
Lyophilized | |
Greater than 95% by SDS-PAGE |