Learn More
Invitrogen™ Human FABP1 (L-FABP) Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75548
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Liver Fatty Acid Binding Protein (human) Recombinant Protein, RP-75548, from Invitrogen.
This product contains a purified 127 amino acid fragment (aa1-127) of human FABP-L protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays. This protein has a molecular weight of ∽18.71 kDa.Specifications
P07148 | |
Control, Western Blot | |
2168 | |
10μg | |
N/A | |
RUO | |
Human FABP1 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
0.41mg/mL | |
tris HCl with 50% glycerol and no preservative | |
Human FABP1 | |
E. Coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Fatty Acid Binding Protein | |
FABP1 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
in vitro research only.