missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human COL9A1 Partial ORF (NP_001842, 24 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001297-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196514

  • £280.00 / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes one of the three alpha chains of type IX collagen, which is a minor (5-20%) collagen component of hyaline cartilage. Type IX collagen is usually found in tissues containing type II collagen, a fibrillar collagen. Studies in knockout mice have shown that synthesis of the alpha 1 chain is essential for assembly of type IX collagen molecules, a heterotrimeric molecule, and that lack of type IX collagen is associated with early onset osteoarthritis. Mutations in this gene are associated with osteoarthritis in humans, with multiple epiphyseal dysplasia, 6, a form of chondrodysplasia, and with Stickler syndrome, a disease characterized by ophthalmic, orofacial, articular, and auditory defects. Two transcript variants that encode different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: AVKRRPRFPVNSNSNGGNELCPKIRIGQDDLPGFDLISQFQVDKAASRRAIQRVVGSATLQVAYKLGNNVDFRIPTRNLYPSGLPEEYSFLTTFRMTGSTLKKNWNIWQ
Specifications

Specifications

NP_001842
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AVKRRPRFPVNSNSNGGNELCPKIRIGQDDLPGFDLISQFQVDKAASRRAIQRVVGSATLQVAYKLGNNVDFRIPTRNLYPSGLPEEYSFLTTFRMTGSTLKKNWNIWQ
RUO
COL9A1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1297
COL9A1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DJ149L1.1.2/EDM6/FLJ40263/MED
COL9A1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers