Learn More
Invitrogen™ Human Alpha-Galactosidase A/GLA ELISA Kit
Human Alpha-Galactosidase A/GLA quantitates human Alpha-Galactosidase A/GLA in serum, plasma, supernatant.
Brand:  Invitrogen™ EH21RB
 View more versions of this product
Product Code. 17184503
  • £540.00 / Pack of 96
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Human Alpha-Galactosidase A/GLA quantitates human Alpha-Galactosidase A/GLA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Alpha-Galactosidase A/GLA.
This gene encodes a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predominantly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; melibiase
45 pg/mL
ELISA
Colorimetric Microplate Reader
GLA
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
45 to 10,000 pg/mL
Biotin
Plasma, Serum, Supernatant
P06280
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
alpha Galactosidase
4 hr 45 min