Learn More
Hoefer™ Gel Plate Rack
Compatible with SE 900 Vertical Electrophoresis System
Brand: Hoefer™ SE914
Code : 04
Additional Details : Weight : 1.00000kg
Description
Holds up to 14 large-format gel cassettes.
Specifications
Up to 14 Large Format Gel Cassette | |
SE 900 Large format Vertical Electrophoresis System | |
Gel Plate Rack |