missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nabertherm High Temperature Tube Furnace for Vertical Operation, RHTV Series
High-quality insulation materials made of vacuum-formed fiber plates enable energy-saving operation and a fast heating time due to low heat storage and heat conductivity. Different gas supply systems allow performance under non-flammable or flammable protective or reactive gases or under vacuum.
£15549.90 - £26509.80
Product Code | Brand | Diameter (Metric) Process Tube | Temperature (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Process Tube | Temperature (Metric) | Price | Quantity & Availability | ||||||
15938260
|
Nabertherm
001484710 |
50 mm | 1600°C | N/A | N/A | N/A | |||||
15958260
|
Nabertherm
001484810 |
50 mm | 1700°C | N/A | N/A | N/A | |||||
15988260
|
Nabertherm
001484910 |
50 mm | 1800°C | N/A | N/A | N/A | |||||
15968260
|
Nabertherm
001484830 |
80 mm | 1700°C | N/A | N/A | N/A | |||||
15998260
|
Nabertherm
001484930 |
80 mm | 1800°C | N/A | N/A | N/A | |||||
15948260
|
Nabertherm
001484750 |
120 mm | 1600°C | N/A | N/A | N/A | |||||
15908270
|
Nabertherm
001484950 |
120 mm | 1800°C | N/A | N/A | N/A | |||||
15978260
|
Nabertherm
001484850 |
120 mm | 1700°C | N/A | N/A | N/A | |||||
Description
- Maximum Temperature: 1600 °C, 1700 °C, or 1800 °C
- MoSi2 heating elements, mounted vertically for easy replacement
- Insulation with vacuum-formed ceramic fiber plates
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Rectangular outer housing with slots for convection cooling
- Tube furnaces RHTV with frame for vertical operation
- Dual shell housing made of sheets of textured stainless steel
- Ceramic working tube made of material C 799 including fiber plugs for operation under air
- Type B thermocouple
- Power unit with low-voltage transformer and thyristor
- Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load and with selectable maximum temperature gradient as tube protection
- Switchgear and control unit separate from furnace in separate floor standing cabinet
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nabertherm High Temperature Tube Furnace for Vertical Operation, RHTV Series