missing translation for 'onlineSavingsMsg'
Learn More

Hexaketocyclohexane octahydrate, 97%, Thermo Scientific Chemicals

£25.30 - £140.00

Chemical Identifiers

CAS 7255-28-9
Molecular Formula C6O6
Molecular Weight (g/mol) 168.06
MDL Number MFCD00001652,MFCD00149074
InChI Key PKRGYJHUXHCUCN-UHFFFAOYSA-N
Synonym cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
PubChem CID 68240
IUPAC Name cyclohexane-1,2,3,4,5,6-hexone
SMILES O=C1C(=O)C(=O)C(=O)C(=O)C1=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10764281
View Documents Promotion Details
Thermo Scientific Acros
156280010
1 g Glass bottle
£25.30
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10031730
View Documents Promotion Details
Thermo Scientific Acros
156280100
10 g Glass bottle
£140.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7255-28-9
168.06
PKRGYJHUXHCUCN-UHFFFAOYSA-N
68240
O=C1C(=O)C(=O)C(=O)C(=O)C1=O
C6O6
MFCD00001652,MFCD00149074
cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
cyclohexane-1,2,3,4,5,6-hexone

Specifications

7255-28-9
Beige to Brown-Gray
97%
MFCD00001652,MFCD00149074
cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
O=C1C(=O)C(=O)C(=O)C(=O)C1=O
168.06
312.17
Powder
90.0°C to 93.0°C
Authentic
C6O6
07, 907
PKRGYJHUXHCUCN-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexone
68240
97%
Hexaketocyclohexane octahydrate, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 208-412-4

Product Identifier
  • Hexaketocyclohexane octahydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only