Learn More
Heptaldehyde, 95%, stabilized, Thermo Scientific Chemicals
£17.10 - £115.00
Chemical Identifiers
CAS | 111-71-7 |
---|---|
Molecular Formula | C7H14O |
Molecular Weight (g/mol) | 114.19 |
MDL Number | MFCD00007028 |
InChI Key | FXHGMKSSBGDXIY-UHFFFAOYSA-N |
Synonym | heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal |
PubChem CID | 8130 |
ChEBI | CHEBI:34787 |
IUPAC Name | heptanal |
SMILES | CCCCCCC=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10497110
|
Thermo Scientific Acros
120320500 |
50 mL | Glass bottle |
£17.10
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10214730
|
Thermo Scientific Acros
120322500 |
250 mL | Glass bottle |
£30.60
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10075903
|
Thermo Scientific Acros
120320010 |
1 L | Glass bottle |
£115.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-71-7 | |
114.19 | |
FXHGMKSSBGDXIY-UHFFFAOYSA-N | |
8130 | |
heptanal |
C7H14O | |
MFCD00007028 | |
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal | |
CHEBI:34787 | |
CCCCCCC=O |
Specifications
111-71-7 | |
Beige-Yellow to White | |
150.0°C | |
Authentic | |
C7H14O | |
1.4115 to 1.4135 | |
01, II, 750 | |
0.82 | |
Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol | |
CCCCCCC=O | |
100ppm hydroquinone | |
8130 | |
0.977 mPa.s (15°C) | |
95% | |
Heptaldehyde, 95% |
-42.0°C | |
0.8200g/mL | |
48°C | |
94% min. (GC) | |
CH3(CH2)5CHO | |
MFCD00007028 | |
15, 4693 | |
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal | |
FXHGMKSSBGDXIY-UHFFFAOYSA-N | |
heptanal | |
114.19 | |
CHEBI:34787 | |
114.19 | |
Powder, Crystals or Chunks |
Safety and Handling
GHS H Statement
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-898-4
missing translation for 'rtecsNumber' : MI6900000
missing translation for 'tsca' : TSCA
RUO – Research Use Only