missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES Free Acid, >99%, Molecular Biology Reagent, MP Biomedicals™

£60.90 - £378.01
Chemical Identifiers
CAS | 7365-45-9 |
---|---|
Molecular Formula | C8H18N2O4S |
Molecular Weight (g/mol) | 238.30 |
MDL Number | MFCD00006158 |
InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
Synonym | N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) |
PubChem CID | 23831 |
ChEBI | CHEBI:42334 |
SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11416561
![]() |
MP Biomedicals
0219482725 |
25 g |
£60.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11426561
![]() |
MP Biomedicals
0219482780 |
100 g |
£123.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11436561
![]() |
MP Biomedicals
0219482790 |
500 g |
£378.01
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Buffering agentChemical Identifiers
7365-45-9 | |
238.30 | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
23831 | |
OCCN1CCN(CCS(O)(=O)=O)CC1 |
C8H18N2O4S | |
MFCD00006158 | |
N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) | |
CHEBI:42334 |
Specifications
7365-45-9 | |
Powder | |
C8H18N2O4S | |
N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
Molecular Biology Reagent |
White | |
>99% | |
MFCD00006158 | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
23831 | |
238.3 | |
HEPES, free acid, Free Acid |
Safety and Handling
missing translation for 'einecsNumber' : 230-907-9
missing translation for 'storageNote' : Store desiccated.
Recommended Storage : Store at Room Temperature (15° to 30°C).
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HEPES Free Acid, >99%, Molecular Biology Reagent, MP Biomedicals™