missing translation for 'onlineSavingsMsg'
Learn More

Solotec Scientific Limited Waysafe™ H14 HEPA Filter For Use With Waysafe™ SA, GP and AS Enclosures

300 SERIES H14 HEPA filter for Waysafe™ SA, AS, AND GP as well as older WS2, WS3, Minicab and mediaway Enclosures. H14 99.995% efficiency in removal of particles that have a size of 0.3 μm. Complete with a 1203.01.00 spring kit

Brand:  Solotec Scientific Limited 300.H14

Code : 04

Additional Details : Weight : 1.00000kg

Product Code. 12707306

  • £550.00 / Each
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Replacement filter, for use with Waysafe™ SA, GP and AS enclosures
Specifications

Specifications

1.0000
Waysafe™ SA, GP and AS Enclosures
1 Filter
32 cm
32 cm
Particulate Filtration
H14 Hepa
13 cm
32 x 32 x 13 cm
H14 Hepa Filter
Pleated Filter Media Bonded in a Purpose Built Galvanised Frame
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers