missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ Hemispheric Bowl

For Use With: Hot Plate Stirrers and Heating Plates

Brand:  Velp Scientifica™ A00000273

Additional Details : Weight : 0.12000kg

Product Code. 15583593

  • £45.65 / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

  • Suitable for Velp Hot plate stirrers and heating plates
  • For use with 135 mm plate hotplates
Specifications

Specifications

50 mL
Fits on 135 mm plate
Hot Plate Stirrers and Heating Plates
Accessory, Bowl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers