Learn More
Hamilton™ Microlab 600 Luer Lock Conversion Kit
This kit converts the Concord CT to a TLL probe.
Brand:  Hamilton™ 58381-01
Code : NEW
Additional Details : Weight : 0.03600kg
Product Code. 15209084
  • £106.00 / Each
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
  • An upgrade for the Concord Convertible Tip Hand Probe
  • Does not include a CT probe
  • Microlab 600 Basic and Advanced
  • Computer controlled
Specifications
Specifications
For liquid handling, and scientific research and laboratory use only
Luer-lok Conversion Kit