Learn More
Hamilton™ Inert Luer Syringe Gas Sampling Valve
Inert gas sampling valve features a lever-actuated on/off valve for storing and transporting samples in any GASTIGHT TLL syringe
Brand:  Hamilton™ 86580
Code : NEW
Additional Details : Weight : 0.02300kg
Product Code. 10373190
  • £115.00 / Each
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
Kel-f hub needles are able to attach to the valve. This valve is pressure tested a 100 psi.
Specifications
Specifications
Storing and transporting samples in any Gastight™ TLL syringe
Inert Gas Sampling Valve