Learn More
Agilent BioTek Half-size Dichroic Mirror
Half-Size Dichroic Mirror, Ex. 475-520, Em. 530-670
Brand: Agilent BioTek 7138525
Additional Details : Weight : 0.80000kg
Description
- Half-Size Dichroic Mirror, Ex. 475-520, Em. 530-670
- For use with Cytation 5, Synergy H1 and Synergy LX
Specifications
Half-size Dichroic Mirror | |
Cytation 3, Cytation 5, Synergy 2, Synergy H1 Multi-Mode Readers | |
Ex. 475-520 nm, Em. 530-670 nm | |
EX 475-520 nm, EM 530-670 nm |