Learn More
Agilent BioTek Half-size Dichroic Mirror
Half-Size Dichroic Mirror, Ex. 400-450, Em. 460-710
Brand: Agilent BioTek 7138455
Additional Details : Weight : 0.01000kg
Description
- Half-Size Dichroic Mirror, Ex. 400-450, Em. 460-710
- For use with Cytation 5, Synergy H1 and Synergy LX
Specifications
Half-size Dichroic Mirror | |
Cytation 3, Cytation 5, Synergy 2, Synergy H1 Multi-Mode Readers | |
400 to 450 nm | |
460 to 710 nm |