missing translation for 'onlineSavingsMsg'
Learn More

Guanosine, 99%, Thermo Scientific Chemicals

Catalog Number p-3736621
37.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
118-00-3
C10H13N5O5
283.24
MFCD00010182
NYHBQMYGNKIUIF-UUOKFMHZSA-N
guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
6802
CHEBI:16750
NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
118-00-3
C10H13N5O5
283.24
MFCD00010182
NYHBQMYGNKIUIF-UUOKFMHZSA-N
guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
6802
CHEBI:16750
NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 250°C
Color White to Yellow
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Beilstein 26, V,18, 81
Merck Index 15, 4601
Specific Rotation Condition −62.00 (20.00°C c=3,in 0.1 N NaOH)
Specific Rotation -62
Solubility Information Solubility in water: slightly soluble. Other solubilities: insoluble in ethanol,ethylether,soluble in acetic acid
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Formula Weight 283.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Guanosine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'