missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Graphite rod, 1.27cm (0.5in) dia x 61cm (24in) long, 99% (metals basis), Thermo Scientific Chemicals
Form: Approx. 130g/pc
£35.00 - £121.00
Chemical Identifiers
CAS | 7782-42-5 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.01 |
MDL Number | MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15425925
|
Thermo Scientific Alfa Aesar
010134.KG |
1 Pc. |
£35.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15435925
|
Thermo Scientific Alfa Aesar
010134.KJ |
5 Pc. |
£121.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-42-5 | |
12.01 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7782-42-5 | |
99% (Metals basis) | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
Rod |
Gray to Black | |
C | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
[C] | |
12.01 | |
CHEBI:27594 | |
Graphite |
Safety and Handling
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only