missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 600 Cellulose General Purpose Filter Paper: Discs

Used in schools and as bench protective paper. Fisherbrand™ Grade 600 Cellulose General Purpose Filter Paper is economical.

Brand:  Fisherbrand™

Code : 47

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11803192

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Applications:
  • Used in hospitals, research laboratories, schools, chemical industries, pharmaceutical, cosmetic, food industries
Specifications

Specifications

Disc
10 to 20 μm
Economical filter Paper used in schools and as bench protective Paper
100/Pk.
100 mm
Cellulose
10 to 20 μm
600
0.15 mm
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers