missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Filter Discs for Qualitative and Technical Analyses, Grade: 3 S/h
Used for analytical and technical purposes.
£8.70 - £49.30
Specifications
Format | Disc |
---|---|
Material | Cotton Linters, Refined Pulp |
Filtration Speed | 55μm/sec. |
For Use With (Application) | For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications |
Grade | 3 S/h |
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | |||||
11770456
|
Sartorius™
FT-3-307-100 |
100 mm |
£10.20
Pack of 50 |
In Stock
Log in to see stock. |
|||||
11780456
|
Sartorius™
FT-3-307-110 |
110 mm |
£10.20
Pack of 50 |
In Stock
Log in to see stock. |
|||||
11710466
|
Sartorius™
FT-3-307-185 |
185 mm |
£25.20
Pack of 50 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
11700466
|
Sartorius™
FT-3-307-150 |
150 mm |
£17.60
Pack of 50 |
In Stock
Log in to see stock. |
|||||
11790456
|
Sartorius™
FT-3-307-125 |
125 mm |
£12.50
Pack of 50 |
In Stock
Log in to see stock. |
|||||
11720466
|
Sartorius™
FT-3-307-240 |
240 mm |
£45.40
Pack of 50 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
11730466
|
Sartorius™
FT-3-307-270 |
270 mm |
£49.30
Pack of 50 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
11740456
|
Sartorius™
FT-3-307-055 |
55 mm |
£8.70
Pack of 50 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
11750456
|
Sartorius™
FT-3-307-070 |
70 mm |
£8.70
Pack of 50 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11760456
|
Sartorius™
FT-3-307-090 |
90 mm |
£8.70
Pack of 50 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
- For routine analyses like clarification, determination of substances
- Discs with a center hole for technical applications
- Made of refined pulp and cotton linters with >95% α-cellulose content
- Smooth surface
- Wet-strengthened
Specifications
Disc | |
55μm/sec. | |
3 S/h | |
0.36 mm |
Cotton Linters, Refined Pulp | |
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications | |
50 Pack | |
Qualitative Filter Paper |