Learn More
SMARCE1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SMARCE1.
Brand: Abnova PAB6459.100ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene is part of the large ATP-dependent chromatin remodeling complex SWI/SNF, which is required for transcriptional activation of genes normally repressed by chromatin. The encoded protein, either alone or when in the SWI/SNF complex, can bind to 4-way junction DNA, which is thought to mimic the topology of DNA as it enters or exits the nucleosome. The protein contains a DNA-binding HMG domain, but disruption of this domain does not abolish the DNA-binding or nucleosome-displacement activities of the SWI/SNF complex. Unlike most of the SWI/SNF complex proteins, this protein has no yeast counterpart. [provided by RefSeq
Sequence: C-PPTDPIPEDEKKESpecifications
SMARCE1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot (0.2-0.6 ug/mL) The optimal working dilution should be determined by the end user. | |
SMARCE1 | |
SMARCE1 | |
A synthetic peptide corresponding to human SMARCE1. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SMARCE1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
BAF57 | |
Goat | |
Antigen affinity purification | |
RUO | |
6605 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only