Learn More
Sartorius Stedim Biotech™ Glass Tube for Perforated Silicone Stopper 75D
Sartorius Stedim Biotech™ Glass Tube for Perforated Silicone Stopper 75D is an integral part of a vacuum flask assembly.
Brand: Sartorius Stedim Biotech™ 1EAQ--0017
Additional Details : Weight : 0.04900kg
Description
- Glass construction
- Fits 75D silicone stopper
- Sturdy enough for vacuum flask operations
Specifications
Glass tube for perforated silicone stopper/vacuum flask assembly | |
Perforated silicone stopper 75 D/ 5L vacuum flask assembly | |
Glass |