missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Distilled Water

High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents

Specifications

PubChemCID
ChEBI
View More Specs

Description

Description

 

  • Suitable for use in cell culture and molecular biology applications: This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
  • Prepared by distillation and filtration (0.1μm), with no added substances
  • Water quality monitored on a routine basis: Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly
  • Produced at a cGMP-compliant facility located in Paisley, Scotland, UK, registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard

 

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.