missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Hybond™ -NX Membranes
Neutral nylon membrane developed specifically for use with low hybridization buffer volumes and high-throughput applications
POA
Specifications
Binding Capacity | Up to 600μg/cm2 |
---|---|
Width (Metric) | 3 m |
Format | Roll |
For Use With (Equipment) | Radioactive probes |
Material | Nylon, Highly Positively Charged |
Description
- Binding capacity for nucleic acids 600μg/cm2
- High tensile strength withstands multiple reprobing
Specifications
Up to 600μg/cm2 | |
Roll | |
Nylon, Highly Positively Charged |
3 m | |
Radioactive probes |