Learn More
Garcin™ Garcin™ Bactinyl Wipers
Clean and disinfect surfaces and instruments with Garcin™ Bactinyl Wipers, which are general purpose wipers for general sanitation purposes.
POA
Specifications
Product Type Wipers
View More Specs
Description
Description
  • Fragranced or odorless
  • Rechargeable dispenser box with reclosable case
  • Active substances include peroxides, quaternary ammoniums, ethanol
  • Mineral-certified impregnation solution
Specifications
Specifications
Wipers