missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | GABARAPL1 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16867783
|
Proteintech
11010-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16857783
|
Proteintech
11010-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The antibody is specific to GABARAPL1, and does not recognize the recombit GABARAP and GABARAPL2.
Increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.Specifications
GABARAPL1 | |
Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q0VGK0, Q8R3R8, Q9H0R8 | |
23710, 57436, 689161 | |
GABARAPL1-Specific Fusion Protein Ag1473 | |
Primary | |
-20°C | |
GABARAPL1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
APG8 LIKE, APG8L, ATG8, ATG8L, GABARAPL1 | |
Gabarapl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |