missing translation for 'onlineSavingsMsg'
Learn More

Labcaire™ Fume cabinet filter AU-1320 exhaust for model no. 250 99.997 % to 0.3μm particles 0.3μm HEPA Aura

Brand:  Labcaire™ AU-1320

Code : 04

Additional Details : Weight : 4.00000kg

Product Code. 12811790

  • £520.46 / Each
Estimated Shipment: 14-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!