missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wesbart™ Freezer Storage Tray
Designed to be placed within a horizontal/ vertical cold store or be free standing
Brand: Wesbart™ WB225A
Code : NEW
Additional Details : Weight : 1.50000kg
Description
Specifications
Plain Storage Tray with High Sides all Around | |
555 x 137 x 13 mm | |
137 mm |
Specify Anodised Aluminum or Stainless Steel | |
555 mm | |
13 mm |