missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wesbart™ Freezer Storage Tray
Designed to be placed within a horizontal/ vertical cold store or be free standing
Brand: Wesbart™ WB1251MTH4
Code : NEW
Additional Details : Weight : 1.50000kg
Description
Specifications
Plain Storage Tray with High Back and Front Sides | |
675 x 133 x 60 mm | |
133 mm |
Specify Anodised Aluminum or Stainless Steel | |
675 mm | |
60 mm |