missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, LC-MS Ultra, Honeywell™ Fluka™

£69.43 - £281.56

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15678310
View Documents Promotion Details
Honeywell-Fluka
14265-1ML
1 mL Ampoule
£69.43
1mL
Please to purchase this item. Need a web account? Register with us today!
15688310
View Documents Promotion Details
Honeywell-Fluka
14265-2ML
2 mL Ampoule
£129.05
2mL
Please to purchase this item. Need a web account? Register with us today!
15671400
View Documents Promotion Details
Honeywell-Fluka
14265-50ML
50 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

64-18-6
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
CH2O2
MFCD00003297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
C(=O)O

Specifications

64-18-6
100.4°C
CH2O2
MFCD00003297
1209246
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.03g/mol
LC-MS Ultra
0.57
1004°F
n20/D 1.370(lit.)
44.8mmHg (20°C)
4°C
Colorless
HCOOH
UN1779
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
C(=O)O
46.025
CHEBI:30751
∼98% (T)
1.22g/mL at 25°C
49°C (120.2°F)
Liquid
1.6 (vs air)
Formic Acid
Videos
Safety and Handling

Safety and Handling

P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233

H226-H302-H314-H331

missing translation for 'dotInformation' : 3(8)/PGII

missing translation for 'einecsNumber' : 200-579-1

missing translation for 'rtecsNumber' : LQ4900000

Product Identifier
  • Formic acid

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : formic acid

SDS
Documents

Documents

Product Certifications