missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde, 37% Solution, BAKER ANALYZED ACS Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
Specifications
CAS | 50-00-0 |
Assay Percent Range | 36.5 to 38.0 % |
Molecular Formula | CH2O |
MDL Number | MFCD00003274 |
Quantity | 500 mL |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
SMILES | C=O |
IUPAC Name | formaldehyde |
Molecular Weight (g/mol) | 30.03 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Signal Word
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'