missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Flexitril™ L27 Nitrile-Coated Chemical/Liquid Protection Gloves
Gloves are flexible and comfortable while also providing protection against harmful chemicals, oils, and greases.
£146.00
Specifications
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, REACH Compliant |
---|---|
Color | Green |
Material | Nitrile |
EN388 Standard | 3121B |
Glove Cuff Style | Gauntlet |
Description
- Gloves combine flexibility, dexterity, and mechanical resistance
- Coating is resistant to chemicals, oils, greases, gasoline, petrol, alcohol, alkali, and salts
Specifications
EN 420:2003 + A1:2009, REACH Compliant | |
Nitrile | |
Gauntlet | |
Lined | |
L27 |
Green | |
3121B | |
Interlock Cotton | |
Chemicals, Oil, Grease, Gasoline, Petrol, Alcohols, Alkalis, and Salts | |
270 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell Flexitril™ L27 Nitrile-Coated Chemical/Liquid Protection Gloves